Assay Details
| Gene Symbol: Thoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024384.2 | NP_077360.2 | 8 - 9 | 826 | 65 | |
| GenBank mRNA | AF275266.1 | - | 1 - 2 | 91 | 65 | |
| BC079149.1 | - | 8 - 9 | 826 | 7128541 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF275266.1 transcripts.
Set Membership: |
|

Thoc6
Genomic Map