Assay Details
| Gene Symbol: LOC688286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247853.2 | XP_006247915.1 | 3 - 4 | 2013 | 97 | |
| XM_006247854.2 | XP_006247916.1 | 5 - 6 | 1267 | 97 | ||
More Information
Set Membership: |
|

LOC688286
Genomic Map