Assay Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030023.1 | NP_001025194.1 | 9 - 10 | 1074 | 152 | |
GenBank mRNA | AY880882.1 | - | 9 - 10 | 1074 | 152 | |
KC795688.1 | - | 10 - 11 | 1107 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
positive regulation of protein phosphorylation
protein phosphorylation
JNK cascade
activation of JUN kinase activity
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
positive regulation of DNA replication
positive regulation of nitric-oxide synthase biosynthetic process
cell growth involved in cardiac muscle cell development
cellular response to mechanical stimulus
cellular response to sorbitol
protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
JUN kinase kinase activity
mitogen-activated protein kinase kinase kinase binding
MAPK cascade
positive regulation of protein phosphorylation
protein phosphorylation
JNK cascade
activation of JUN kinase activity
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
positive regulation of DNA replication
positive regulation of nitric-oxide synthase biosynthetic process
cell growth involved in cardiac muscle cell development
cellular response to mechanical stimulus
cellular response to sorbitol
protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
JUN kinase kinase activity
mitogen-activated protein kinase kinase kinase binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151718 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P462132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138525 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P462132 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151718 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151718 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P462132 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P462132 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151718 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19143 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P462132 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60724 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80399 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74277 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7432 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66757 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10743675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743671 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10743668 | - | - |
Rat Genome 230 2.0 Array | 1377621_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377621_at | - | - |
Rat Genome 230 2.0 Array | 1383701_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383701_at | - | - |
Affymetrix HT Rat Focus Array | 1383701_at | - | - |
Affymetrix HT Rat Focus Array | 1383701_PM_at | - | - |
Rat Expression Set 230 | 1383701_PM_at | - | - |
Rat Expression Set 230 | 1383701_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10743668 | - | - |
Rat Genome U34 Set | rc_AI058348_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369256 | - | - |
