Assay Details
| Gene Symbol: LOC499602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025039.2 | NP_001020210.2 | 4 - 5 | 348 | 69 | |
| XM_006232310.2 | XP_006232372.1 | 4 - 5 | 568 | 69 | ||
| XM_008760940.1 | XP_008759162.1 | 3 - 4 | 207 | 69 | ||
| GenBank mRNA | CK358190.1 | - | 4 - 5 | 406 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC499602
Genomic Map