Assay Details
Gene Symbol: LOC499602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025039.2 | NP_001020210.2 | 10 - 11 | 874 | 111 | |
XM_006232310.2 | XP_006232372.1 | 9 - 10 | 1004 | 111 | ||
XM_006232311.2 | XP_006232373.1 | 6 - 7 | 541 | 111 | ||
XM_008760940.1 | XP_008759162.1 | 9 - 10 | 733 | 111 | ||
GenBank mRNA | BC098046.1 | - | 6 - 7 | 513 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
