Assay Details
| Gene Symbol: Fabp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134614.1 | NP_001128086.1 | 2 - 3 | 327 | 91 | |
| XM_006232145.2 | XP_006232207.1 | 2 - 3 | 931 | 91 | ||
| XM_006232146.2 | XP_006232208.1 | 2 - 3 | 460 | 91 | ||
| XM_006232147.2 | XP_006232209.1 | 1 - 2 | 128 | 91 | ||
| XM_006232148.2 | XP_006232210.1 | 1 - 2 | 128 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp12
Genomic Map