Assay Details
| Gene Symbol: Ckm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012530.2 | NP_036662.1 | 3 - 4 | 442 | 127 | |
| GenBank mRNA | DN931989.1 | - | 3 - 4 | 441 | 127 | |
| FQ214594.1 | - | 3 - 4 | 418 | 127 | ||
| FQ214750.1 | - | 3 - 4 | 418 | 127 | ||
| FQ214825.1 | - | 2 - 3 | 239 | 127 | ||
| FQ214931.1 | - | 3 - 4 | 419 | 127 | ||
| FQ215369.1 | - | 2 - 3 | 316 | 127 | ||
| FQ215410.1 | - | 3 - 4 | 418 | 127 | ||
| FQ215927.1 | - | 2 - 3 | 271 | 127 | ||
| FQ216321.1 | - | 2 - 3 | 270 | 127 | ||
| FQ216507.1 | - | 3 - 4 | 413 | 127 | ||
| FQ217421.1 | - | 2 - 3 | 283 | 127 | ||
| FQ224848.1 | - | 2 - 3 | 269 | 127 | ||
| FQ224918.1 | - | 3 - 4 | 419 | 127 | ||
| M10140.1 | - | 3 - 4 | 415 | 127 | ||
| M14864.1 | - | 1 - 2 | 88 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ckm
Genomic Map