Assay Details
| Gene Symbol: Gucy1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017090.2 | NP_058786.2 | 3 - 4 | 452 | 95 | |
| XM_006232518.2 | XP_006232580.1 | 4 - 5 | 1044 | 95 | ||
| XM_008761107.1 | XP_008759329.1 | 2 - 3 | 1075 | 95 | ||
| XM_008761108.1 | XP_008759330.1 | 3 - 4 | 818 | 95 | ||
| GenBank mRNA | AB096020.1 | - | 2 - 3 | 315 | 95 | |
| BC085746.1 | - | 3 - 4 | 452 | 7190419 | 95 | |
| M57405.1 | - | 3 - 4 | 771 | 95 | ||
| U60835.1 | - | 3 - 4 | 771 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy1a3
Genomic Map