Assay Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191978.1 | NP_001178907.1 | 2 - 3 | 313 | 121 | |
| GenBank mRNA | FQ227184.1 | - | 2 - 3 | 382 | 121 | |
| FQ227324.1 | - | 2 - 3 | 382 | 121 | ||
| GU357482.1 | - | 2 - 3 | 163 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clec1b
Genomic Map