Assay Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191978.1 | NP_001178907.1 | 4 - 5 | 590 | 94 | |
| XM_006237088.2 | XP_006237150.1 | 4 - 5 | 2166 | 94 | ||
| XM_006237089.2 | XP_006237151.1 | 3 - 4 | 911 | 94 | ||
| XM_006237090.2 | XP_006237152.1 | 3 - 4 | 2068 | 94 | ||
| GenBank mRNA | FQ227324.1 | - | 4 - 5 | 659 | 94 | |
| GU357482.1 | - | 4 - 5 | 440 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clec1b
Genomic Map