Assay Details
Gene Symbol: Fam21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199207.1 | NP_954677.1 | 11 - 12 | 1060 | 133 | |
XM_006237147.2 | XP_006237209.1 | 11 - 12 | 1084 | 133 | ||
XM_006237149.2 | XP_006237211.1 | 11 - 12 | 1084 | 133 | ||
GenBank mRNA | AY257251.1 | - | 11 - 12 | 1060 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
