Assay Details
Gene Symbol: Ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031979.3 | NP_114185.3 | 3 - 4 | 517 | 82 | |
GenBank mRNA | D28557.1 | - | 3 - 4 | 381 | 82 | |
U22893.1 | - | 3 - 4 | 520 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
