Assay Details
Gene Symbol: Klrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012745.2 | NP_036877.1 | 1 - 2 | 314 | 113 | |
XM_006237078.2 | XP_006237140.1 | 1 - 2 | 432 | 113 | ||
GenBank mRNA | AF009133.1 | - | 1 - 2 | 71 | 113 | |
BC086393.1 | - | 1 - 2 | 314 | 7303354 | 113 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF009133.1,BC086393.1,NM_012745.2,XM_006237078.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
