Assay Details
| Gene Symbol: Fgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131908.1 | NP_571983.1 | 1 - 2 | 350 | 118 | |
| GenBank mRNA | AB079674.1 | - | 1 - 2 | 350 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgf6
Genomic Map