Assay Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106626.1 | NP_001100096.1 | 1 - 2 | 206 | 74 | |
| XM_006237595.2 | XP_006237657.1 | 3 - 4 | 922 | 74 | ||
| XM_008763376.1 | XP_008761598.1 | 3 - 4 | 902 | 74 | ||
| XM_008763377.1 | XP_008761599.1 | 3 - 4 | 923 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription corepressor activity
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription corepressor activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P312371 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167971 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14101 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P312371 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P312371 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14101 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14101 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P312371 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19424 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P251840 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49520 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10859482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10859487 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10859478 | - | - |
| Rat Genome 230 2.0 Array | 1371996_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1385728_at | - | - |
| Rat Genome 230 2.0 Array | 1371996_at | - | - |
| Rat Genome 230 2.0 Array | 1381994_at | - | - |
| Rat Genome 230 2.0 Array | 1385728_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381994_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400014369.rn.1 | TC0400001985.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0400014370.rn.1 | TC0400001985.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10859478 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22359451 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367081 | - | - |

Aebp2
Genomic Map