Assay Details
Gene Symbol: Qrich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247833.2 | XP_006247895.2 | 17 - 18 | 5739 | 62 | |
XM_008768433.1 | XP_008766655.1 | 18 - 19 | 5709 | 62 | ||
XM_008768434.1 | XP_008766656.1 | 17 - 18 | 5409 | 62 | ||
XM_008768435.1 | XP_008766657.1 | 16 - 17 | 5199 | 62 | ||
XM_008768436.1 | XP_008766658.1 | 17 - 18 | 5139 | 62 | ||
XM_008768437.1 | XP_008766659.1 | 15 - 16 | 3968 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
