Assay Details
| Gene Symbol: LOC102555947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246839.2 | XP_006246901.1 | 1 - 2 | 188 | 92 | |
| Gene Symbol: Cntrob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134645.1 | NP_001128117.1 | 10 - 11 | 1659 | 92 | |
| XM_006246701.2 | XP_006246763.1 | 11 - 12 | 2033 | 92 | ||
| XM_006246702.2 | XP_006246764.1 | 10 - 11 | 2307 | 92 | ||
| XM_008767825.1 | XP_008766047.1 | 10 - 11 | 1367 | 92 | ||
| GenBank mRNA | EF532342.1 | - | 10 - 11 | 1659 | 92 | |
| EF532343.1 | - | 10 - 11 | 1711 | 92 | ||
| EF532344.1 | - | 10 - 11 | 1718 | 92 | ||
| EF532345.1 | - | 10 - 11 | 1711 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC102555947
Genomic Map