Assay Details
Gene Symbol: Hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247774.2 | XP_006247836.1 | 8 - 9 | 1136 | 70 | |
XM_213523.8 | XP_213523.6 | 8 - 9 | 1132 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
