Assay Details
| Gene Symbol: Kif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053377.1 | NP_445829.1 | 4 - 5 | 624 | 63 | |
| XM_006246347.2 | XP_006246409.1 | 4 - 5 | 662 | 63 | ||
| XM_006246351.2 | XP_006246413.1 | 4 - 5 | 661 | 63 | ||
| XM_008767716.1 | XP_008765938.1 | 4 - 5 | 812 | 63 | ||
| XM_008767717.1 | XP_008765939.1 | 4 - 5 | 661 | 63 | ||
| GenBank mRNA | AM180763.1 | - | 2 - 3 | 216 | 63 | |
| CV116151.1 | - | 2 - 3 | 99 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif3a
Genomic Map