Assay Details
Gene Symbol: Tmem256 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170549.1 | NP_001164020.1 | 3 - 4 | 227 | 85 | |
XM_008767803.1 | XP_008766025.1 | 3 - 4 | 205 | 85 | ||
GenBank mRNA | BF549068.1 | - | 3 - 4 | 233 | 85 | |
CB719779.1 | - | 1 - 2 | 85 | |||
FQ212389.1 | - | 3 - 4 | 214 | 85 | ||
FQ223049.1 | - | 3 - 4 | 211 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
