Assay Details
| Gene Symbol: Ndel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133320.1 | NP_579854.1 | 5 - 6 | 575 | 71 | |
| XM_006246571.2 | XP_006246633.1 | 5 - 6 | 984 | 71 | ||
| XM_006246572.2 | XP_006246634.1 | 5 - 6 | 977 | 71 | ||
| XM_006246573.2 | XP_006246635.1 | 5 - 6 | 725 | 71 | ||
| XM_006246574.2 | XP_006246636.1 | 5 - 6 | 610 | 71 | ||
| XM_006246575.2 | XP_006246637.1 | 5 - 6 | 1088 | 71 | ||
| XM_006246576.2 | XP_006246638.1 | 5 - 6 | 978 | 71 | ||
| XM_006246578.2 | XP_006246640.1 | 5 - 6 | 724 | 71 | ||
| GenBank mRNA | AY008298.1 | - | 5 - 6 | 575 | 71 | |
| BC070909.1 | - | 5 - 6 | 665 | 7103650 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndel1
Genomic Map