Assay Details
| Gene Symbol: Clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022393.2 | NP_071788.2 | 3 - 4 | 483 | 85 | |
| XM_006246803.2 | XP_006246865.1 | 3 - 4 | 521 | 85 | ||
| XM_008767867.1 | XP_008766089.1 | 3 - 4 | 521 | 85 | ||
| XM_008767868.1 | XP_008766090.1 | 3 - 4 | 365 | 85 | ||
| XM_008767869.1 | XP_008766091.1 | 3 - 4 | 362 | 85 | ||
| GenBank mRNA | BC091151.1 | - | 3 - 4 | 483 | 7375234 | 85 |
| FQ213042.1 | - | 3 - 4 | 355 | 85 | ||
| FQ221204.1 | - | 3 - 4 | 372 | 85 | ||
| FQ228437.1 | - | 3 - 4 | 341 | 85 | ||
| J05495.1 | - | 3 - 4 | 343 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clec10a
Genomic Map