Assay Details
| Gene Symbol: Cd300lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001061779.4 | XP_001061779.2 | 7 - 8 | 933 | 121 | |
| XM_002727812.3 | XP_002727858.1 | 6 - 7 | 801 | 121 | ||
| XM_003750936.3 | XP_003750984.1 | 7 - 8 | 1185 | 121 | ||
| XM_003750937.3 | XP_003750985.1 | 6 - 7 | 1053 | 121 | ||
More Information
Set Membership: |
|

Cd300lg
Genomic Map