Assay Details
| Gene Symbol: Dhrs7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271597.1 | NP_001258526.1 | 4 - 5 | 631 | 66 | |
| NM_001271598.1 | NP_001258527.1 | 4 - 5 | 631 | 66 | ||
| GenBank mRNA | FN805189.1 | - | 4 - 5 | 586 | 66 | |
| FQ214628.1 | - | 4 - 5 | 613 | 66 | ||
| FQ214964.1 | - | 4 - 5 | 611 | 66 | ||
| FQ215164.1 | - | 4 - 5 | 612 | 66 | ||
| FQ216481.1 | - | 4 - 5 | 613 | 66 | ||
| FQ216936.1 | - | 4 - 5 | 612 | 66 | ||
| FQ224852.1 | - | 4 - 5 | 612 | 66 | ||
| FQ224951.1 | - | 4 - 5 | 611 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhrs7c
Genomic Map