Assay Details
Gene Symbol: Srp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108840.1 | NP_001102310.1 | 1500 | 65 | ||
XM_008768381.1 | XP_008766603.1 | 5 - 6 | 2214 | 65 | ||
GenBank mRNA | BC166787.1 | - | 1397 | 7935892 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
