Assay Details
Gene Symbol: Rpl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246606.2 | XP_006246668.1 | 1 - 2 | 195 | 56 | |
GenBank mRNA | FQ209942.1 | - | 1 - 2 | 41 | 56 | |
FQ210265.1 | - | 1 - 2 | 41 | 56 | ||
FQ210305.1 | - | 1 - 2 | 42 | 56 | ||
FQ214577.1 | - | 1 - 2 | 41 | 56 | ||
FQ218083.1 | - | 1 - 2 | 41 | 56 | ||
FQ218115.1 | - | 1 - 2 | 40 | 56 | ||
FQ220102.1 | - | 41 | 56 | |||
FQ221223.1 | - | 1 - 2 | 43 | 56 | ||
FQ221302.1 | - | 1 - 2 | 41 | 56 | ||
FQ221303.1 | - | 1 - 2 | 41 | 56 | ||
FQ221304.1 | - | 1 - 2 | 41 | 56 | ||
FQ221369.1 | - | 1 - 2 | 41 | 56 | ||
FQ221596.1 | - | 1 - 2 | 36 | 56 | ||
FQ221712.1 | - | 1 - 2 | 41 | 56 | ||
FQ221762.1 | - | 1 - 2 | 41 | 56 | ||
FQ222089.1 | - | 1 - 2 | 41 | 56 | ||
FQ222135.1 | - | 1 - 2 | 41 | 56 | ||
FQ222176.1 | - | 1 - 2 | 41 | 56 | ||
FQ222323.1 | - | 1 - 2 | 35 | 56 | ||
FQ222676.1 | - | 1 - 2 | 41 | 56 | ||
FQ222872.1 | - | 1 - 2 | 41 | 56 | ||
FQ222877.1 | - | 1 - 2 | 35 | 56 | ||
FQ222953.1 | - | 1 - 2 | 41 | 56 | ||
FQ223076.1 | - | 1 - 2 | 42 | 56 | ||
FQ223161.1 | - | 1 - 2 | 41 | 56 | ||
FQ223442.1 | - | 1 - 2 | 41 | 56 | ||
FQ224611.1 | - | 1 - 2 | 41 | 56 | ||
FQ228453.1 | - | 1 - 2 | 40 | 56 | ||
FQ228549.1 | - | 1 - 2 | 35 | 56 | ||
FQ228592.1 | - | 1 - 2 | 35 | 56 | ||
FQ228636.1 | - | 1 - 2 | 41 | 56 | ||
FQ228935.1 | - | 1 - 2 | 42 | 56 | ||
X14671.1 | - | 1 - 2 | 36 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the X14671.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
