Assay Details
| Gene Symbol: Trappc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039378.1 | NP_001034467.1 | 1 - 2 | 37 | 97 | |
| GenBank mRNA | AY903234.1 | - | 1 - 2 | 37 | 97 | |
| AY903235.1 | - | 1 - 2 | 37 | 97 | ||
| AY903236.1 | - | 1 - 2 | 37 | 97 | ||
| AY903237.1 | - | 1 - 2 | 37 | 97 | ||
| AY903238.1 | - | 1 - 2 | 37 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY903234.1, AY903235.1, AY903236.1, AY903237.1, AY903238.1, NM_001039378.1 transcripts.
Set Membership: |
|

Trappc1
Genomic Map