Assay Details
Gene Symbol: Gnb2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130734.2 | NP_570090.2 | 2 - 3 | 418 | 59 | |
GenBank mRNA | BC063809.1 | - | 2 - 3 | 393 | 6919982 | 59 |
FQ212638.1 | - | 2 - 3 | 389 | 59 | ||
FQ213067.1 | - | 2 - 3 | 389 | 59 | ||
FQ213571.1 | - | 2 - 3 | 390 | 59 | ||
FQ215527.1 | - | 2 - 3 | 389 | 59 | ||
FQ215812.1 | - | 2 - 3 | 389 | 59 | ||
FQ216167.1 | - | 2 - 3 | 389 | 59 | ||
FQ216212.1 | - | 2 - 3 | 388 | 59 | ||
FQ218475.1 | - | 2 - 3 | 419 | 59 | ||
FQ218650.1 | - | 2 - 3 | 393 | 59 | ||
FQ219434.1 | - | 2 - 3 | 389 | 59 | ||
FQ219492.1 | - | 2 - 3 | 390 | 59 | ||
FQ219693.1 | - | 2 - 3 | 389 | 59 | ||
FQ219729.1 | - | 2 - 3 | 389 | 59 | ||
FQ219878.1 | - | 2 - 3 | 387 | 59 | ||
FQ219900.1 | - | 2 - 3 | 389 | 59 | ||
FQ220124.1 | - | 2 - 3 | 389 | 59 | ||
FQ220151.1 | - | 2 - 3 | 389 | 59 | ||
FQ220277.1 | - | 2 - 3 | 389 | 59 | ||
FQ220314.1 | - | 2 - 3 | 389 | 59 | ||
FQ220391.1 | - | 2 - 3 | 389 | 59 | ||
FQ220419.1 | - | 2 - 3 | 389 | 59 | ||
FQ220428.1 | - | 2 - 3 | 393 | 59 | ||
FQ220430.1 | - | 2 - 3 | 388 | 59 | ||
FQ220472.1 | - | 2 - 3 | 389 | 59 | ||
FQ220494.1 | - | 2 - 3 | 389 | 59 | ||
FQ220585.1 | - | 2 - 3 | 389 | 59 | ||
FQ220598.1 | - | 2 - 3 | 393 | 59 | ||
FQ220697.1 | - | 2 - 3 | 393 | 59 | ||
FQ220819.1 | - | 2 - 3 | 390 | 59 | ||
FQ220976.1 | - | 2 - 3 | 396 | 59 | ||
FQ221075.1 | - | 2 - 3 | 389 | 59 | ||
FQ221140.1 | - | 2 - 3 | 396 | 59 | ||
FQ221286.1 | - | 2 - 3 | 393 | 59 | ||
FQ222502.1 | - | 2 - 3 | 389 | 59 | ||
FQ223855.1 | - | 2 - 3 | 388 | 59 | ||
FQ224952.1 | - | 2 - 3 | 389 | 59 | ||
FQ225047.1 | - | 2 - 3 | 393 | 59 | ||
FQ225312.1 | - | 2 - 3 | 388 | 59 | ||
FQ225511.1 | - | 2 - 3 | 389 | 59 | ||
FQ225843.1 | - | 2 - 3 | 419 | 59 | ||
FQ226802.1 | - | 2 - 3 | 389 | 59 | ||
FQ228007.1 | - | 2 - 3 | 393 | 59 | ||
FQ229157.1 | - | 2 - 3 | 388 | 59 | ||
FQ229247.1 | - | 2 - 3 | 389 | 59 | ||
FQ229458.1 | - | 2 - 3 | 386 | 59 | ||
FQ229569.1 | - | 2 - 3 | 390 | 59 | ||
FQ229616.1 | - | 2 - 3 | 389 | 59 | ||
FQ229633.1 | - | 2 - 3 | 389 | 59 | ||
FQ229689.1 | - | 2 - 3 | 389 | 59 | ||
FQ229758.1 | - | 2 - 3 | 389 | 59 | ||
FQ229908.1 | - | 2 - 3 | 390 | 59 | ||
FQ229911.1 | - | 2 - 3 | 389 | 59 | ||
FQ229916.1 | - | 2 - 3 | 387 | 59 | ||
FQ229923.1 | - | 2 - 3 | 388 | 59 | ||
FQ230003.1 | - | 2 - 3 | 387 | 59 | ||
FQ230018.1 | - | 2 - 3 | 397 | 59 | ||
FQ230117.1 | - | 2 - 3 | 389 | 59 | ||
FQ230328.1 | - | 2 - 3 | 390 | 59 | ||
FQ233024.1 | - | 2 - 3 | 389 | 59 | ||
FQ234000.1 | - | 2 - 3 | 395 | 59 | ||
FQ234673.1 | - | 2 - 3 | 391 | 59 | ||
FQ235255.1 | - | 2 - 3 | 389 | 59 | ||
U03390.1 | - | 2 - 3 | 381 | 59 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
