Assay Details
| Gene Symbol: Dis3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127483.1 | NP_001120955.1 | 19 - 20 | 2756 | 74 | |
| XM_006252391.2 | XP_006252453.1 | 20 - 21 | 2617 | 74 | ||
| XM_008770869.1 | XP_008769091.1 | 19 - 20 | 2691 | 74 | ||
| GenBank mRNA | BC166824.1 | - | 19 - 20 | 2756 | 8370691 | 74 |
| FQ210067.1 | - | 19 - 20 | 2768 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dis3
Genomic Map