Assay Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012043.1 | NP_001012043.1 | 11 - 12 | 2133 | 75 | |
GenBank mRNA | BC081929.1 | - | 11 - 12 | 2133 | 7113366 | 75 |
FQ226413.1 | - | 1 - 2 | 763 | 75 | ||
FQ232347.1 | - | 1 - 2 | 763 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128506 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21574 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21574 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21574 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21574 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19698 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77893 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91731 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71224 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66837 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3876 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103967 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96238 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40060 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101106 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10781763 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781766 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781761 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781756 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781760 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781757 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781765 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781762 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781759 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781758 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10781752 | - | - |
Rat Genome 230 2.0 Array | 1398401_at | - | - |
Rat Genome 230 2.0 Array | 1398401_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398401_at | - | - |
Affymetrix HT Rat Focus Array | 1398401_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500006424.rn.1 | TC1500000938.rn.1 | RTA TC |
Rat Expression Set 230 | 1398401_PM_at | - | - |
Rat Expression Set 230 | 1398401_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10781752 | - | - |
Rat Genome U34 Set | rc_AI058808_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21944487 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376936 | - | - |
