Assay Details
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107281.1 | NP_001100751.1 | 5 - 6 | 1028 | 61 | |
XM_006252408.2 | XP_006252470.1 | 5 - 6 | 979 | 61 | ||
XM_006252409.2 | XP_006252471.1 | 5 - 6 | 974 | 61 | ||
XM_006252410.2 | XP_006252472.1 | 5 - 6 | 1045 | 61 | ||
XM_006252413.2 | XP_006252475.1 | 4 - 5 | 827 | 61 | ||
XM_006252415.2 | XP_006252477.1 | 4 - 5 | 824 | 61 | ||
XM_008770870.1 | XP_008769092.1 | 4 - 5 | 2781 | 61 | ||
XM_008770871.1 | XP_008769093.1 | 6 - 7 | 991 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
