Assay Details
| Gene Symbol: Scel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108388.1 | NP_001101858.1 | 30 - 31 | 2090 | 66 | |
| XM_006252428.2 | XP_006252490.1 | 31 - 32 | 2172 | 66 | ||
| XM_008770882.1 | XP_008769104.1 | 31 - 32 | 2154 | 66 | ||
| XM_008770883.1 | XP_008769105.1 | 30 - 31 | 2115 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P150879 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P150884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721679 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155453 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150879 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150884 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150884 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150879 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P150879 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P150884 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P189793 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71822 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39826 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6418 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10781915 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781922 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781905 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781921 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781898 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781923 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781918 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781920 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781909 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781897 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781910 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781925 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781926 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781924 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781907 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781912 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781904 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781917 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781908 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781895 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10781890 | - | - |
| Rat Genome 230 2.0 Array | 1385099_at | - | - |
| Rat Genome 230 2.0 Array | 1385099_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385099_at | - | - |
| Affymetrix HT Rat Focus Array | 1385099_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500006822.rn.1 | TC1500001018.rn.1 | RTA TC |
| Rat Expression Set 230 | 1385099_at | - | - |
| Rat Expression Set 230 | 1385099_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781890 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22361658 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355076 | - | - |

Scel
Genomic Map