Assay Details
| Gene Symbol: N4bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033893.1 | NP_001029065.1 | 1 - 2 | 324 | 109 | |
| XM_008767681.1 | XP_008765903.1 | 547 | 109 | |||
| GenBank mRNA | DQ176639.1 | - | 1 - 2 | 324 | 109 | |
| Gene Symbol: LOC103689949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246253.2 | XP_006246315.1 | 2 - 3 | 547 | 109 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

N4bp3
Genomic Map