Assay Details
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768202.1 | XP_008766424.1 | 8 - 9 | 1725 | 83 | |
XM_008768203.1 | XP_008766425.1 | 8 - 9 | 1726 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
