Assay Details
| Gene Symbol: Krt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173128.1 | NP_775151.1 | 3 - 4 | 815 | 64 | |
| GenBank mRNA | M63665.1 | - | 3 - 4 | 815 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt20
Genomic Map