Assay Details
Gene Symbol: Abca17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031637.1 | NP_001026807.1 | 26 - 27 | 4517 | 102 | |
XM_006245927.2 | XP_006245989.1 | 27 - 28 | 5625 | 102 | ||
XM_006245928.2 | XP_006245990.1 | 27 - 28 | 5609 | 102 | ||
XM_008767554.1 | XP_008765776.1 | 27 - 28 | 6239 | 102 | ||
XM_008767555.1 | XP_008765777.1 | 26 - 27 | 4612 | 102 | ||
XM_008767556.1 | XP_008765778.1 | 26 - 27 | 4498 | 102 | ||
XM_008767557.1 | XP_008765779.1 | 26 - 27 | 4531 | 102 | ||
GenBank mRNA | AB196699.1 | - | 26 - 27 | 4517 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P125705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67394 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125705 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P125705 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P125705 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P497239 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80947 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74077 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69464 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50496 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78307 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66781 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50760 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10741053 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741061 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741046 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741062 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741073 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741063 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741066 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741047 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741057 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741064 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741055 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741069 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741058 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741052 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741072 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741056 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741071 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741048 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741070 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741045 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741051 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741050 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741060 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741065 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741068 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741067 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741049 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741059 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10741054 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10741044 | - | - |
Affymetrix Rat Clariom S Assay | PSR1000021111.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021116.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021112.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021115.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021117.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021113.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021114.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000021118.rn.1 | TC1000002386.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10741044 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22027301 | - | - |
ABI Rat Genome Survey Microarray | 22399826 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374113 | - | - |
