Assay Details
| Gene Symbol: RGD1310166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247061.2 | XP_006247123.1 | 2 - 3 | 221 | 107 | |
| XM_220804.7 | XP_220804.5 | 2 - 3 | 252 | 107 | ||
More Information
Set Membership: |
|

RGD1310166
Genomic Map