Assay Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247940.2 | XP_006248002.1 | 5 - 6 | 667 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc26a11| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247940.2 | XP_006248002.1 | 5 - 6 | 667 | 65 | |
Set Membership: |
|