Assay Details
| Gene Symbol: Rasl10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191648.1 | NP_001178577.1 | 2 - 3 | 402 | 114 | |
| Gene Symbol: LOC103694863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773809.1 | XP_008772031.1 | 2 - 3 | 868 | 114 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rasl10b
Genomic Map