Assay Details
| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005876.1 | NP_001005876.1 | 2 - 3 | 302 | 107 | |
| GenBank mRNA | BC079311.1 | - | 2 - 3 | 302 | 7134794 | 107 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hn1
Genomic Map