Assay Details
| Gene Symbol: Zfp213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245862.2 | XP_006245924.1 | 4 - 5 | 998 | 101 | |
| XM_006245863.2 | XP_006245925.1 | 4 - 5 | 1534 | 101 | ||
More Information
Set Membership: |
|

Zfp213
Genomic Map