Assay Details
Gene Symbol: Zfp213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105764.1 | NP_001099234.1 | 3 - 4 | 902 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P147358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24737 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147358 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147358 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P147358 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P547561 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10740829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740830 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10740815 | - | - |
Rat Genome 230 2.0 Array | 1378688_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377575_PM_s_at | - | - |
Rat Genome 230 2.0 Array | 1377575_s_at | - | - |
Rat Genome 230 2.0 Array | 1377285_at | - | - |
Rat Genome 230 2.0 Array | 1372892_at | - | - |
Rat Genome 230 2.0 Array | 1390361_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390361_at | - | - |
Rat Genome 230 2.0 Array | 1392042_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372892_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377285_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392042_at | - | - |
Rat Genome 230 2.0 Array | 1378688_at | - | - |
Affymetrix HT Rat Focus Array | 1390361_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372892_at | - | - |
Affymetrix HT Rat Focus Array | 1377575_s_at | - | - |
Affymetrix HT Rat Focus Array | 1377575_PM_s_at | - | - |
Affymetrix HT Rat Focus Array | 1390361_at | - | - |
Affymetrix HT Rat Focus Array | 1392042_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392042_at | - | - |
Affymetrix HT Rat Focus Array | 1372892_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000020718.rn.1 | TC1000002353.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020719.rn.1 | TC1000002353.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020721.rn.1 | TC1000002353.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000020720.rn.1 | TC1000002353.rn.1 | RTA TC |
Rat Expression Set 230 | 1372892_PM_at | - | - |
Rat Expression Set 230 | 1377285_at | - | - |
Rat Expression Set 230 | 1377285_PM_at | - | - |
Rat Expression Set 230 | 1390361_PM_at | - | - |
Rat Expression Set 230 | 1372892_at | - | - |
Rat Expression Set 230 | 1390361_at | - | - |
Rat Expression Set 230 | 1392042_at | - | - |
Rat Expression Set 230 | 1378688_PM_at | - | - |
Rat Expression Set 230 | 1378688_at | - | - |
Rat Expression Set 230 | 1377575_s_at | - | - |
Rat Expression Set 230 | 1392042_PM_at | - | - |
Rat Expression Set 230 | 1377575_PM_s_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10740815 | - | - |
Rat Genome U34 Set | rc_AI007755_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358616 | - | - |
