Assay Details
| Gene Symbol: Alox15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153301.2 | NP_695213.1 | 6 - 7 | 881 | 60 | |
| XM_008767779.1 | XP_008766001.1 | 5 - 6 | 742 | 60 | ||
| GenBank mRNA | AF415240.1 | - | 6 - 7 | 889 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Alox15b
Genomic Map