Assay Details
| Gene Symbol: Zfp354a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246227.2 | XP_006246289.1 | 1 - 2 | 1412 | 98 | |
| XM_008767660.1 | XP_008765882.1 | 1 - 2 | 1412 | 98 | ||
| XM_008767661.1 | XP_008765883.1 | 1 - 2 | 1412 | 98 | ||
| GenBank mRNA | BC078777.1 | - | 1 - 2 | 102 | 7127868 | 98 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078777.1, XM_006246227.2, XM_008767660.1, XM_008767661.1 transcripts.
Set Membership: |
|

Zfp354a
Genomic Map