Assay Details
Gene Symbol: Dvl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172056.1 | NP_001165527.1 | 9 - 10 | 1066 | 122 | |
XM_006246715.2 | XP_006246777.1 | 9 - 10 | 1286 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
