Assay Details
Gene Symbol: Spata20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199402.1 | NP_955434.1 | 14 - 15 | 2172 | 79 | |
XM_006247200.2 | XP_006247262.1 | 14 - 15 | 2198 | 79 | ||
GenBank mRNA | AY438568.1 | - | 14 - 15 | 2172 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
