Assay Details
| Gene Symbol: Sugt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013051.1 | NP_001013069.1 | 11 - 12 | 776 | 76 | |
| GenBank mRNA | BC158724.1 | - | 11 - 12 | 776 | 9090831 | 76 |
| FQ221379.1 | - | 11 - 12 | 802 | 76 | ||
| FQ227975.1 | - | 11 - 12 | 894 | 76 | ||
| FQ232174.1 | - | 11 - 12 | 894 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P039522 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P030634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156785 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030634 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039522 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039522 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030634 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P039522 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P030634 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P729705 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1006279 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59330 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9776 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38829 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10781702 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781703 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781701 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781705 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781704 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781700 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10781699 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10785444 | - | - |
| Rat Genome 230 2.0 Array | 1372627_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372627_at | - | - |
| Rat Genome 230 2.0 Array | 1383364_at | - | - |
| Rat Genome 230 2.0 Array | 1383364_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372627_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372627_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500006301.rn.1 | TC1500000910.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372627_at | - | - |
| Rat Expression Set 230 | 1372627_PM_at | - | - |
| Rat Expression Set 230 | 1383364_PM_at | - | - |
| Rat Expression Set 230 | 1383364_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10781699 | - | - |
| Rat Genome U34 Set | rc_AA901343_at | - | - |
| Rat Genome U34 Set | rc_AI101283_at | - | - |
| Rat Genome U34 Set | rc_AI136494_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350869 | - | - |

Sugt1
Genomic Map