Assay Details
| Gene Symbol: Esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106051.3 | NP_001099521.1 | 3 - 4 | 213 | 95 | |
| NM_001270865.1 | NP_001257794.1 | 3 - 4 | 295 | 95 | ||
| NM_001270866.1 | NP_001257795.1 | 3 - 4 | 213 | 95 | ||
| GenBank mRNA | BC158790.1 | - | 3 - 4 | 204 | 7308742 | 95 |
| FQ213565.1 | - | 3 - 4 | 213 | 95 | ||
| FQ216373.1 | - | 3 - 4 | 297 | 95 | ||
| FQ218750.1 | - | 3 - 4 | 95 | |||
| FQ229329.1 | - | 3 - 4 | 187 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esd
Genomic Map