Assay Details
| Gene Symbol: Supt7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108010.2 | NP_001101480.1 | 3 - 4 | 1008 | 89 | |
| GenBank mRNA | BC168944.1 | - | 3 - 4 | 1008 | 8371512 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Supt7l
Genomic Map