Assay Details
Gene Symbol: Rpl36al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031105.2 | NP_112367.1 | 1 - 2 | 80 | ||
GenBank mRNA | BC058142.1 | - | 6919511 | 80 | ||
FQ218934.1 | - | 1 - 2 | 63 | 80 |
Gene Symbol: LOC100912182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751738.3 | XP_003751786.1 | 1 - 2 | 89 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC058142.1,NM_031105.2,XM_003751738.3 transcripts.
This assay is in the 5'UTR region of the BC058142.1,NM_031105.2,XM_003751738.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
